2-Bromo-4,6-difluorobenzoic acid ≥95%

Supplier: AOBChem USA

Warning

25256-1G 25256-25G 25256-500MG 25256-5G
76899-932EA 195.21 USD
76899-932 76899-934 76899-936 76899-938
2-Bromo-4,6-difluorobenzoic acid ≥95%
2-Bromo-4,6-difluorobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₃BrF₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD23713104
CAS Number: 651026-99-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR