4-Chloro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 2-Chloro-5-formylphenylboronic acid pinacol ester

25166-25G 25166-5G
76899-884EA 559.1 USD
76899-884 76899-886
4-Chloro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde ≥97% research grade
4-Chloro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde
Formula: C₁₃H₁₆BClO₃
MW: 266.52 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18731013
CAS Number: 1112209-14-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR