2-Chloro-4-(trifluoromethyl)benzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Chloro-4-(trifluoromethyl)benzylalcohol

25151-25G 25151-5G
76899-874EA 254.62 USD
76899-874 76899-876
2-Chloro-4-(trifluoromethyl)benzyl alcohol ≥97%
2-Chloro-4-(trifluoromethyl)benzyl alcohol

Precaución: for Research Use Only

Formula: C₈H₆ClF₃O
MW: 210.58 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04972757
CAS Number: 56456-51-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR