2-Chloro-6-(trifluoromethyl)phenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

25116-1G 25116-25G 25116-500MG 25116-5G
76899-860EA 135.8 USD
76899-860 76899-862 76899-864 76899-866
2-Chloro-6-(trifluoromethyl)phenylboronic acid pinacol ester ≥97%
2-Chloro-6-(trifluoromethyl)phenylboronic acid pinacol ester
Formula: C₁₃H₁₅BClF₃O₂
MW: 306.516 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18756702
CAS Number: 1451391-09-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR