2,5-Difluoro-4-(Methylsulfanyl)Phenylboronic Acid ≥97%
Supplier: AOBChem USA
|
Warning
|
25060-1G
25060-25G
25060-500MG
25060-5G
76899-824EA
155.96
USD
76899-824
76899-826
76899-828
76899-830
2,5-Difluoro-4-(Methylsulfanyl)Phenylboronic Acid ≥97%
2,5-Difluoro-4-(Methylsulfanyl)Phenylboronic Acid
Formula:
C₇H₇BF₂O₂S MW: 204.002 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD22628252 CAS Number: 1451392-37-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...