(4-Fluoro-2,6-dimethylphenyl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2,6-Dimethyl-4-fluorophenylboronic acid

25042-1G 25042-25G 25042-500MG 25042-5G
76899-812EA 171.87 USD
76899-812 76899-814 76899-816 76899-818
(4-Fluoro-2,6-dimethylphenyl)boronic acid ≥97%
(4-Fluoro-2,6-dimethylphenyl)boronic acid
Formula: C₈H₁₀BFO₂
MW: 167.97 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD23712929
CAS Number: 1392512-54-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR