2-Fluoro-4-methoxypyridine ≥97% research grade

Supplier: AOBChem USA

Warning

25028-1G 25028-25G 25028-5G
76899-800EA 76.39 USD
76899-800 76899-802 76899-804
2-Fluoro-4-methoxypyridine ≥97% research grade
2-Fluoro-4-methoxypyridine
Formula: C₆H₆FNO
Storage Temperature: Ambient
MDL Number: MFCD12828014
CAS Number: 175965-83-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR