2-Fluoro-3-phenylpyridine-5-boronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

25010-1G 25010-500MG 25010-5G
76899-786EA 485.9 USD
76899-786 76899-788 76899-790
2-Fluoro-3-phenylpyridine-5-boronic acid ≥97% research grade
2-Fluoro-3-phenylpyridine-5-boronic acid
Formula: C₁₁H₉BFNO₂
Storage Temperature: Ambient
MDL Number: MFCD28015431
CAS Number: 2121513-18-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR