2-Fluoro-3-methoxybenzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Fluoro-3-(hydroxymethyl)anisole

25005-25G 25005-5G
76899-776EA 595.17 USD
76899-776 76899-778
2-Fluoro-3-methoxybenzyl alcohol ≥97%
2-Fluoro-3-methoxybenzyl alcohol
Formula: C₈H₉FO₂
MW: 156.154 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09743596
CAS Number: 178974-59-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR