2,5-Dichloropyridin-3-ol ≥97% research grade

Supplier: AOBChem USA

Warning

24949-1G 24949-500MG 24949-5G
76899-766EA 471.04 USD
76899-766 76899-768 76899-770
2,5-Dichloropyridin-3-ol ≥97% research grade
2,5-Dichloropyridin-3-ol
Formula: C₅H₃Cl₂NO
Storage Temperature: Ambient
MDL Number: MFCD13185537
CAS Number: 53335-73-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR