2-Methoxy-4,6-bis(trifluoromethyl)benzoic acid ≥95%

Supplier: AOBChem USA

Warning

24931-25G 24931-5G
76899-754EA 595.17 USD
76899-754 76899-756
2-Methoxy-4,6-bis(trifluoromethyl)benzoic acid ≥95%
2-Methoxy-4,6-bis(trifluoromethyl)benzoic acid
Formula: C₁₀H₆F₆O₃
Storage Temperature: Ambient
MDL Number: MFCD00728738
CAS Number: 180134-15-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR