2-Bromo-5-(trifluoromethoxy)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

24906-1G 24906-25G 24906-500MG 24906-5G
76899-738EA 161.26 USD
76899-738 76899-740 76899-742 76899-744
2-Bromo-5-(trifluoromethoxy)benzaldehyde ≥97%
2-Bromo-5-(trifluoromethoxy)benzaldehyde
Formula: C₈H₄BrF₃O₂
MW: 269.01 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07368722
CAS Number: 505084-61-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR