2-Chloro-4-cyclopropylpyridine ≥97%

Supplier: AOBChem USA

Warning

24884-1G 24884-25G 24884-500MG 24884-5G
76899-708EA 215.37 USD
76899-708 76899-710 76899-712 76899-714
2-Chloro-4-cyclopropylpyridine ≥97%
2-Chloro-4-cyclopropylpyridine
Formula: C₈H₈ClN
Storage Temperature: Ambient
MDL Number: MFCD12032792
CAS Number: 168975-75-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR