2-Choro-3,5-difluorophenylboronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: (2-Chloro-3,5-difluorophenyl)boronic acid

24867-1G 24867-25G 24867-500MG 24867-5G
76899-694EA 290.69 USD
76899-694 76899-696 76899-698 76899-700
2-Choro-3,5-difluorophenylboronic acid ≥97% research grade
2-Choro-3,5-difluorophenylboronic acid
Formula: C₆H₄BClF₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD18415643
CAS Number: 1451393-37-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR