2-Methoxy-4-pyridineboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Methoxy-4-pyridylboronic acid Pinacol ester, 2-Methoxy-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 2-(2-Methoxy-4-pyridyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-Methoxypyridine-4-boronic acid pinacol ester

24852-1G 24852-25G 24852-5G
76899-684EA 62.6 USD
76899-684 76899-686 76899-688
2-Methoxy-4-pyridineboronic acid pinacol ester ≥97%
2-Methoxy-4-pyridineboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₂H₁₈BNO₃
Storage Temperature: Refrigerator
MDL Number: MFCD07368867
CAS Number: 408502-23-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR