2-Fluoro-4-formyl-6-methoxyphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

24848-1G 24848-25G 24848-5G
76899-670EA 96.55 USD
76899-670 76899-672 76899-674
2-Fluoro-4-formyl-6-methoxyphenylboronic acid ≥97%
2-Fluoro-4-formyl-6-methoxyphenylboronic acid
Formula: C₈H₈BFO₄
Storage Temperature: Refrigerator
MDL Number: MFCD22628230
CAS Number: 1451392-10-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR