2-Cyclopropylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

24818-1G 24818-25G 24818-500MG 24818-5G
76899-654EA 215.37 USD
76899-654 76899-656 76899-658 76899-660
2-Cyclopropylbenzaldehyde ≥97%
2-Cyclopropylbenzaldehyde
Formula: C₁₀H₁₀O
Storage Temperature: Refrigerator
MDL Number: MFCD00480888
CAS Number: 20034-51-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR