2-Cyclopropylpyridine-3-carbaldehyde ≥97%

Supplier: AOBChem USA

Warning

24771-1G 24771-500MG 24771-5G 24771-25G
76899-642EA 468.92 USD
76899-642 76899-644 76899-646 77635-558
2-Cyclopropylpyridine-3-carbaldehyde ≥97%
2-Cyclopropylpyridine-3-carbaldehyde

Precaución: for Research Use Only

Formula: C₉H₉NO
MW: 147.174 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18255582
CAS Number: 921760-70-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR