2-Fluoro-6-methoxy-4-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

24708-1G 24708-25G 24708-500MG 24708-5G
76899-604EA 257.8 USD
76899-604 76899-606 76899-608 76899-610
2-Fluoro-6-methoxy-4-methylbenzaldehyde ≥97%
2-Fluoro-6-methoxy-4-methylbenzaldehyde
Formula: C₉H₉FO₂
MW: 168.165 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22628207
CAS Number: 1362295-77-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR