2-Chloro-5-cyclopropylpyridine-3-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

24689-1G 24689-500MG 24689-5G 24689-25G
76899-592EA 549.55 USD
76899-592 76899-594 76899-596 77658-194
2-Chloro-5-cyclopropylpyridine-3-carboxylic acid ≥97%
2-Chloro-5-cyclopropylpyridine-3-carboxylic acid
Formula: C₉H₈ClNO₂
MW: 197.618 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20660025
CAS Number: 1035690-32-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR