2-Bromo-1,4-difluoro-5-(methylthio)benzene ≥97%

Supplier: AOBChem USA

Warning

24642-25G 24642-5G
76899-576EA 453.01 USD
76899-576 76899-578
2-Bromo-1,4-difluoro-5-(methylthio)benzene ≥97%
2-Bromo-1,4-difluoro-5-(methylthio)benzene
Formula: C₇H₅BrF₂S
MW: 239.08 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22628202
CAS Number: 1383968-48-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR