2-Isopropoxy-4-methylphenylboronic acid ≥95%

Supplier: AOBChem USA

Warning

24528-1G 24528-25G 24528-500MG 24528-5G
76899-532EA 201.58 USD
76899-532 76899-534 76899-536 76899-538
2-Isopropoxy-4-methylphenylboronic acid ≥95%
2-Isopropoxy-4-methylphenylboronic acid
Formula: C₁₀H₁₅BO₃
Storage Temperature: Ambient
MDL Number: MFCD22543720
CAS Number: 221111-03-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR