2-Ethoxy-4-methylphenylboronic acid ≥95% research grade

Supplier: AOBChem USA

Warning

24524-1G 24524-25G 24524-500MG 24524-5G
76899-516EA 284.33 USD
76899-516 76899-518 76899-520 76899-522
2-Ethoxy-4-methylphenylboronic acid ≥95% research grade
2-Ethoxy-4-methylphenylboronic acid
Formula: C₉H₁₃BO₃
Storage Temperature: Ambient
MDL Number: MFCD22543718
CAS Number: 1451391-71-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR