2-Chloro-3,5-dimethylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

24485-1G 24485-25G 24485-500MG 24485-5G
76899-500EA 320.4 USD
76899-500 76899-502 76899-504 76899-506
2-Chloro-3,5-dimethylbenzaldehyde ≥97%
2-Chloro-3,5-dimethylbenzaldehyde
Formula: C₉H₉ClO
MW: 168.62 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11846007
CAS Number: 125340-12-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR