2-Bromo-4-chloro-1-isopropoxybenzene ≥97%

Supplier: AOBChem USA

Warning

24455-1G 24455-25G 24455-5G
76899-478EA 78.51 USD
76899-478 76899-480 76899-482
2-Bromo-4-chloro-1-isopropoxybenzene ≥97%
2-Bromo-4-chloro-1-isopropoxybenzene
Formula: C₉H₁₀BrClO
MW: 249.532 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00070758
CAS Number: 201849-19-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR