2-Bromo-4-fluoro-5-iodobenzotrifluoride ≥95%

Supplier: AOBChem USA

Warning

24399-1G 24399-25G 24399-500MG 24399-5G
76899-438EA 171.87 USD
76899-438 76899-440 76899-442 76899-444
2-Bromo-4-fluoro-5-iodobenzotrifluoride ≥95%
2-Bromo-4-fluoro-5-iodobenzotrifluoride
Formula: C₇H₂BrF₄I
MW: 368.893 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22543695
CAS Number: 1394291-55-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR