2-Chloro-1-iodo-4-isopropoxybenzene ≥97%

Supplier: AOBChem USA

Warning

24315-1G 24315-25G 24315-500MG 24315-5G
76899-406EA 201.58 USD
76899-406 76899-408 76899-410 76899-412
2-Chloro-1-iodo-4-isopropoxybenzene ≥97%
2-Chloro-1-iodo-4-isopropoxybenzene
Formula: C₉H₁₀ClIO
Storage Temperature: Ambient
MDL Number: MFCD22628191
CAS Number: 1369830-17-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR