2-Bromo-3-(difluoromethoxy)pyridine ≥97%

Supplier: AOBChem USA

Warning

24178-1G 24178-25G 24178-500MG 24178-5G
76899-374EA 124.13 USD
76899-374 76899-376 76899-378 76899-380
2-Bromo-3-(difluoromethoxy)pyridine ≥97%
2-Bromo-3-(difluoromethoxy)pyridine

Precaución: for Research Use Only

Formula: C₆H₄BrF₂NO
MW: 224.003 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD13185840
CAS Number: 947249-27-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR