2-[(5-Bromo-2-pyridinyl)oxy]oxy]-ethanol ≥97%

Supplier: AOBChem USA

Warning

24070-1G 24070-25G 24070-500MG 24070-5G
76899-352EA 224.92 USD
76899-352 76899-354 76899-356 76899-358
2-[(5-Bromo-2-pyridinyl)oxy]oxy]-ethanol ≥97%
2-[(5-Bromo-2-pyridinyl)oxy]oxy]-ethanol
Formula: C₇H₈BrNO₂
MW: 218.048 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13190751
CAS Number: 212961-31-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR