2-Fluoro-4-iodo-5-(methoxymethoxy)pyridine ≥97%

Supplier: AOBChem USA

Warning

24028-1G 24028-25G 24028-500MG 24028-5G
76899-324EA 147.47 USD
76899-324 76899-326 76899-328 76899-330
2-Fluoro-4-iodo-5-(methoxymethoxy)pyridine ≥97%
2-Fluoro-4-iodo-5-(methoxymethoxy)pyridine

Precaución: for Research Use Only

Formula: C₇H₇FINO₂
MW: 283.039 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22123918
CAS Number: 1034467-27-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR