2,5-Dibromopyridin-4-ol ≥97%

Supplier: AOBChem USA

Warning

23983-1G 23983-25G 23983-500MG 23983-5G
76899-276EA 228.1 USD
76899-276 76899-278 76899-280 76899-282
2,5-Dibromopyridin-4-ol ≥97%
2,5-Dibromopyridin-4-ol
Formula: C₅H₃Br₂NO
MW: 252.891 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11100682
CAS Number: 1033203-55-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR