2-Phenoxyisonicotinic acid ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 2-Phenoxypyridine-4-carboxylic acid

23979-1G 23979-25G 23979-5G
76899-262EA 114.58 USD
76899-262 76899-264 76899-266
2-Phenoxyisonicotinic acid ≥95%
2-Phenoxyisonicotinic acid
Formula: C₁₂H₉NO₃
MW: 215.21 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09414712
CAS Number: 51362-08-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR