2,4-Difluorothioanisole ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2,4-Difluoro-1-(methylsulphanyl)benzene

23910-25G 23910-5G
76899-244EA 346.92 USD
76899-244 76899-246
2,4-Difluorothioanisole ≥97%
2,4-Difluorothioanisole

Precaución: for Research Use Only

Formula: C₇H₆F₂S
MW: 160.19 g/mol
Storage Temperature: Ambient
CAS Number: 130922-40-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR