2-Methoxy-6-(3-nitrophenyl)pyridine ≥95%

Supplier: AOBChem USA

Warning

23891-1G 23891-25G 23891-500MG 23891-5G
76899-234EA 203.7 USD
76899-234 76899-236 76899-238 76899-240
2-Methoxy-6-(3-nitrophenyl)pyridine ≥95%
2-Methoxy-6-(3-nitrophenyl)pyridine
Formula: C₁₂H₁₀N₂O₃
MW: 230.219 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19981559
CAS Number: 1313359-04-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR