5-Iodo-N,N-dimethylpyridin-2-amine ≥97%
Supplier: AOBChem USA
|
Warning
|
Synonyms:
2-Dimethylamino-5-iodopyridine
23812-1G
23812-25G
23812-500MG
23812-5G
76899-210EA
142.17
USD
76899-210
76899-212
76899-214
76899-216
5-Iodo-N,N-dimethylpyridin-2-amine ≥97%
5-Iodo-N,N-dimethylpyridin-2-amine
Formula:
C₇H₉IN₂ MW: 248.06 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD18207670 CAS Number: 380381-36-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...