2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-4-(trifluoromethyl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Formyl-5-(trifluoromethyl)phenylboronic acid pinacol ester

23754-1G 23754-25G 23754-500MG 23754-5G
76899-194EA 350.1 USD
76899-194 76899-196 76899-198 76899-200
2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-4-(trifluoromethyl)benzaldehyde ≥97%
2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-4-(trifluoromethyl)benzaldehyde
Formula: C₁₄H₁₆BF₃O₃
MW: 300.08 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18757662
CAS Number: 1416721-24-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR