2,6-Dimethoxyisonicotinic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2,6-Dimethoxypyridine-4-carboxylic acid

23693-1G 23693-25G 23693-500MG 23693-5G
76899-164EA 171.87 USD
76899-164 76899-166 76899-168 76899-170
2,6-Dimethoxyisonicotinic acid ≥97%
2,6-Dimethoxyisonicotinic acid
Formula: C₈H₉NO₄
MW: 183.17 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18257556
CAS Number: 6274-82-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR