2-Bromo-4-fluoro-5-methoxybenzoic acid ≥97% research grade

Supplier: AOBChem USA

Warning

23664-1G 23664-500MG 23664-5G 23664-25G
76899-150EA 530.45 USD
76899-150 76899-152 76899-154 77654-338
2-Bromo-4-fluoro-5-methoxybenzoic acid ≥97% research grade
2-Bromo-4-fluoro-5-methoxybenzoic acid
Formula: C₈H₆BrFO₃
MW: 249.034 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD13193242
CAS Number: 1007455-21-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR