2-Bromo-6-chlorothioanisole ≥97%

Supplier: AOBChem USA

Warning

23656-25G 23656-5G
76899-144EA 453.01 USD
76899-144 76899-146
2-Bromo-6-chlorothioanisole ≥97%
2-Bromo-6-chlorothioanisole
Formula: C₇H₆BrClS
Storage Temperature: Ambient
MDL Number: MFCD22056776
CAS Number: 1370025-57-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR