6-Phenyl-3-formylpyridine ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Phenyl-5-pyridinecarboxaldehyde, 6-Phenylnicotinaldehyde

23564-1G 23564-25G 23564-500MG 23564-5G
76899-130EA 126.25 USD
76899-130 76899-132 76899-134 76899-136
6-Phenyl-3-formylpyridine ≥97%
6-Phenyl-3-formylpyridine

Precaución: for Research Use Only

Formula: C₁₂H₉NO
MW: 183.21
Storage Temperature: Ambient
MDL Number: MFCD03086130
CAS Number: 63056-20-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR