2-Fluoro-4-iodo-5-methoxypyridine ≥97%

Supplier: AOBChem USA

Warning

23505-1G 23505-25G 23505-500MG 23505-5G
76899-088EA 260.99 USD
76899-088 76899-090 76899-092 76899-094
2-Fluoro-4-iodo-5-methoxypyridine ≥97%
2-Fluoro-4-iodo-5-methoxypyridine
Formula: C₆H₅FINO
Storage Temperature: Refrigerator
MDL Number: MFCD16611137
CAS Number: 1227516-18-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR