2-Fluoro-4-pyridone ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Fluoropyridin-4-ol, 2-Fluoro-4-hydroxypyridine

23460-1G 23460-25G 23460-5G
76899-074EA 81.69 USD
76899-074 76899-076 76899-078
2-Fluoro-4-pyridone ≥97%
2-Fluoro-4-pyridone

Precaución: for Research Use Only

Formula: C₅H₄FNO
Storage Temperature: Refrigerator
MDL Number: MFCD04114159
CAS Number: 22282-69-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR