2-Fluoro-5-methoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

23403-25G 23403-5G
76899-038EA 492.26 USD
76899-038 76899-040
2-Fluoro-5-methoxybenzoic acid ≥97%
2-Fluoro-5-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₈H₇FO₃
Storage Temperature: Ambient
MDL Number: MFCD00272566
CAS Number: 367-83-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR