5-Bromo-2-methyl-2H-indazole ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Methyl-5-bromo-2H-indazole

23389-25G 23389-5G
76899-026EA 225.98 USD
76899-026 76899-028
5-Bromo-2-methyl-2H-indazole ≥97%
5-Bromo-2-methyl-2H-indazole

Precaución: for Research Use Only

Formula: C₈H₇BrN₂
Storage Temperature: Ambient
MDL Number: MFCD09870047
CAS Number: 465529-56-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR