2-Chloro-1-iodo-4-(trifluoromethoxy)benzene ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 2-Chloro-4-(trifluoromethoxy)iodobenzene, 3-Chloro-4-iodophenyl trifluoromethyl ether, 3-Chloro-4-iodo-α,α,α-trifluoroanisole

23312-25G 23312-5G
76899-014EA 310.85 USD
76899-014 76899-016
2-Chloro-1-iodo-4-(trifluoromethoxy)benzene ≥95%
2-Chloro-1-iodo-4-(trifluoromethoxy)benzene

Precaución: for Research Use Only

Formula: C₇H₃ClF₃IO
MW: 322.451 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08458089
CAS Number: 345226-19-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR