2,6-Dichloro-3-(trifluoromethyl)benzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

23039-25G 23039-5G
76898-992EA 418 USD
76898-992 76898-994
2,6-Dichloro-3-(trifluoromethyl)benzyl alcohol ≥97%
2,6-Dichloro-3-(trifluoromethyl)benzyl alcohol

Precaución: for Research Use Only

Formula: C₈H₅Cl₂F₃O
Storage Temperature: Ambient
MDL Number: MFCD11519348
CAS Number: 1092461-04-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR