2-Bromo-5-iodo-1,3-dimethylbenzene ≥97%

Supplier: AOBChem USA

Warning

23027-25G 23027-5G
76898-982EA 183.54 USD
76898-982 76898-984
2-Bromo-5-iodo-1,3-dimethylbenzene ≥97%
2-Bromo-5-iodo-1,3-dimethylbenzene
Formula: C₈H₈BrI
Storage Temperature: Ambient
MDL Number: MFCD07780650
CAS Number: 689260-53-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR