2,4-Difluoro-3-isopropoxyphenylboronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

23021-1G 23021-25G 23021-500MG 23021-5G
76898-970EA 129.43 USD
76898-970 76898-972 76898-974 76898-976
2,4-Difluoro-3-isopropoxyphenylboronic acid ≥97% research grade
2,4-Difluoro-3-isopropoxyphenylboronic acid
Formula: C₉H₁₁BF₂O₃
Storage Temperature: Ambient
MDL Number: MFCD19981529
CAS Number: 1451390-95-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR