2-Fluoro-4,5-dimethoxyphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

22999-1G 22999-25G 22999-500MG 22999-5G
76898-956EA 231.28 USD
76898-956 76898-958 76898-960 76898-962
2-Fluoro-4,5-dimethoxyphenylboronic acid ≥97%
2-Fluoro-4,5-dimethoxyphenylboronic acid
Formula: C₈H₁₀BFO₄
Storage Temperature: Ambient
MDL Number: MFCD19440154
CAS Number: 900175-07-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR