2-Fluoro-4-(trifluoromethyl)phenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Fluoro-4-trifluoromethylphenylboronic acid pinacol ester, 2-(2-Fluoro-4-(trifluoromethyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-Fluoro-4-(trifluoromethyl)benzeneboronic acid pinacol ester

22876-1G 22876-25G 22876-5G
76898-930EA 62.6 USD
76898-930 76898-932 76898-934
2-Fluoro-4-(trifluoromethyl)phenylboronic acid pinacol ester ≥97%
2-Fluoro-4-(trifluoromethyl)phenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₅BF₄O₂
Storage Temperature: Ambient
MDL Number: MFCD08458187
CAS Number: 1073353-68-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR